Brij® C20

Supplier: Thermo Scientific Chemicals

Synonyms: Polyoxyethylene(20) cetyl ether

34429-0025 34429-5000
TS34429-0025EA 280 USD
TS34429-0025 TS34429-5000
Brij® C20
Brij® C20
Formula: C₁₆H₃₃(OCH₂CH₂)₂₀OH
MW: 1123.5 g/mol
Melting Pt: 35.4…39.6 °C
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00080892
CAS Number: 9004-95-9
EINECS: 500-014-1

Order Now

Specification Test Results

Appearance White Pellets
Infrared spectrum Conforms
Water ≤1 %
Additional info remark: without antioxidant
Impurity ≤1 ppm (Free ethyleneoxide)
Dioxane ≤10 ppm
Acid value ≤1 mg KOH/g
Hydroxyl value 45 to 60 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR