Polyethylene glycol, M.W. ≈ M.W. 600,000
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly(ethylene oxide), Macrogol, PEO, PEG, Carbowax
17861-5000
17861-2500
TS17861-5000EA
183
USD
TS17861-5000
TS17861-2500
Polyethylene glycol, M.W. ≈ M.W. 600,000
Polyethylene glycol
Formula:
H(OCH₂CH₂)nOH Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Specification Test Results
Appearance | White Granular powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...