ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt hydrate (NADPH-Na4 hydrate, reduced form) 96%, extra pure

Supplier: Thermo Scientific Chemicals

32874-0010 32874-2500
TS32874-0010EA 1550 USD
TS32874-0010 TS32874-2500
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt hydrate (NADPH-Na4 hydrate, reduced form) 96%, extra pure
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt hydrate (NADPH-Na4 hydrate, reduced form)
Formula: C₂₁H₂₆N₇Na₄O₁₇P₃·xH₂O
Storage Temperature: Freezer
MDL Number: MFCD10567216
CAS Number: 2646-71-1
EINECS: 220-163-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
HPLC ≥95 %
Sodium (Na) 10 to 12 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR