Quinine sulphate dihydrate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulphuric acid dihydrate, (-)-Quinine sulphate dihydrate

16371-0100 16371-0500
TS16371-0100EA 114 USD
TS16371-0100 TS16371-0500
Quinine sulphate dihydrate 99+%
Quinine sulphate dihydrate
Formula: C₄₀H₅₀N₄O₈S·2H₂O
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00150790
CAS Number: 6119-70-6
EINECS: 212-359-2
Merck Index: 12,03822

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals or needles
Infrared spectrum Conforms
Water 4.0 to 5.5 % (K.F.)
Residue after ignition ≤0.1 %
Specific optical rotation -235° to -245° (20°C, 589 nm) (c=2, 0.1N HCl) (on anhydrous basis)
Titration with HClO4 99.0 to 101.0 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR