Zinc chloride 98+%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
19684-0010
19684-0025
AC19684-0050
AC19684-0100
TS19684-0010EA
130
USD
TS19684-0010
TS19684-0025
TS19684-0050
TS19684-0100
Zinc chloride 98+%, extra pure
Zinc chloride
Formula:
ZnCl₂ Boiling Pt: 732 °C (760 mmHg) Melting Pt: 293 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 13,10185 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals and/or chunks |
Titration Complexometric | ≥98.0 % |
Cadmium (Cd) | ≤10 ppm |
Copper (Cu) | ≤10 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤10 ppm |
Manganese (Mn) | ≤30 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...