1-Octanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium octane-1-sulphonate
39669-1000
AC39669-0250
TS39669-1000EA
1029
USD
TS39669-1000
TS39669-0250
1-Octanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
1-Octanesulphonic acid sodium salt
Formula:
C₈H₁₇NaO₃S MW: 216.28 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007544 CAS Number: 5324-84-5 EINECS: 226-195-4 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (150°C) |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.4 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...