n-Hexane 95+%, extra_pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexyl hydride, Hexanen, Hexane
19736-0025
19736-0050
197360010
197360250
TS19736-0025EA
203
USD
TS19736-0025
TS19736-0050
TS197360010
TS197360250
n-Hexane 95+%, extra_pure
n-Hexane
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 69 Melting Pt: -94.3 Density: 0.659 g/cm³ Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3 II Merck Index: 13,04712 |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.05 % (K.F.) |
GC | ≥95.0 % |
Color scale | ≤20 APHA |
Residue after evaporation | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...