Hexane (mixture of isomers) 99% ACS for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

41155-0025 411555000
TS41155-0025EA 171 USD
TS41155-0025 TS411555000
Hexane (mixture of isomers) 99% ACS for spectroscopy
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
GC ≥98.5 % Sum of 5 isomers + total Hexanes + Methylcyclopentane
UV at 195 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g
Thiophene passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR