Hydrazine hydrate 55% in aqueous solution (∼35% w/w N₂H₄)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrazinium hydroxide, Hydrazine monohydrate

29681-5000 29681-0025 29681-1000
TS29681-5000EA 131 USD
TS29681-5000 TS29681-0025 TS29681-1000
Hydrazine hydrate 55% in aqueous solution (∼35% w/w N₂H₄)
Hydrazine hydrate
Formula: NH₂NH₂·xH₂O
MW: 32.05 g/mol (anhy)
Melting Pt: –65 °C
Density: 1.023 g/cm³
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149931
CAS Number: 10217-52-4
EINECS: 206-114-9
UN: 3293
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless Solution
Refractive index 1.3860 to 1.3900 (20°C, 589 nm)
Density (20°C) 1.021 to 1.025 g/ml
Titration with HCl 54.5 to 55.5 % (N2H4.xH2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR