4-Methylmorpholine N-oxide hydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: NMO hydrate

20834-1000 20834-0250
TS20834-1000EA 323 USD
TS20834-1000 TS20834-0250
4-Methylmorpholine N-oxide hydrate 97%
4-Methylmorpholine N-oxide hydrate
Formula: C₅H₁₁NO₂.xH₂O
MW: 117.147 g/mol (anhy)
Melting Pt: 70…75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005947
CAS Number: 80913-66-2
EINECS: 231-391-8

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤15 % (K.F.)
Titration with HClO4 ≥96.0 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR