2-Mercaptopyridine-N-oxide sodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Hydroxy-2-pyridinethione, sodium salt hydrate, 2-Pyridinethiol-1-oxide, sodium salt hydrate, Sodium omadine hydrate

25538-0250 25538-0050
TS25538-0250EA 224 USD
TS25538-0250 TS25538-0050
2-Mercaptopyridine-N-oxide sodium salt hydrate 98%
2-Mercaptopyridine-N-oxide sodium salt hydrate
Formula: C₅H₄NNaOS.nH₂O
MW: 149.14 g/mol (anhy)
Melting Pt: >260 °C
Storage Temperature: Ambient
MDL Number: MFCD00151244
CAS Number: 207511-13-5
EINECS: 223-296-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Off-white to light yellow Powder or granules and/or chunks
Infrared spectrum Conforms
Loss on drying ≤12 %
Titration with HClO4 ≥97.5 % (On dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR