Chromosulfuric acid

Supplier: Thermo Scientific Chemicals
Danger

29551-0010 295510025
TS29551-0010EA 107 USD
TS29551-0010 TS295510025
Chromosulfuric acid
Chromosulfuric acid
Formula: Cr₂K₂O₇·H₂O₄S
Boiling Pt: 330 °C (760 mmHg)
Density: 1.84 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011368
CAS Number: 65272-71-1
UN: 2240
ADR: 8,I

Order Now

Specification Test Results

Appearance Red-brown to dark brown Liquid
Assay ≥1.3 % CrO3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR