Bis(trifluoromethanesulfonyl)imide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
lithium bis(trifluoromethanesulphonimide), 1,1,1-Trifluoro-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N,N-Bis(trifluoromethylsulphonyl)amine
38760-0250
38760-0010
38760-0050
TS38760-0250EA
443
USD
TS38760-0250
TS38760-0010
TS38760-0050
Bis(trifluoromethanesulfonyl)imide 99%
Bis(trifluoromethanesulfonyl)imide
Formula:
C₂HF₆NO₄S₂ MW: 281.143 g/mol Boiling Pt: 90…91 °C (760 mmHg) Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00214154 CAS Number: 82113-65-3 UN: 3261 ADR: 8 I |
Specification Test Results
Appearance | Colorless to brown Crystals |
Infrared spectrum | Conforms |
Melting point | 50 to 56 °C |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...