Di-n-butylmagnesium 0.5 M in heptane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Magnesium,butyl(1-methylpropyl)-, dibutyl-magnesiu, Magnesium, dibutyl-

37777-1000 AC37777-8000
TS37777-1000EA 148 USD
TS37777-1000 TS37777-8000
Di-n-butylmagnesium 0.5 M in heptane, AcroSeal™
Di-n-butylmagnesium
Formula: C₈H₁₈Mg
Melting Pt: –71 °C
Density: 0.73 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00015225
CAS Number: 1191-47-5
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless Solution
Titration 9.1 to 10.8 % (on Mg basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR