Rhodamine WT 20% in water
Supplier: Thermo Scientific Chemicals
44697-0010
44697-1000
TS44697-0010EA
445
USD
TS44697-0010
TS44697-1000
Rhodamine WT 20% in water
Rhodamine WT
Formula:
C₂₉H₂₉ClN₂Na₂O₅ Density: 1.16 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD01728973 CAS Number: 37299-86-8 |
Specification Test Results
Appearance | Dark red Liquid |
UV Lambda max | 553 to 557 nm |
Strength | 19.5 to 21 % (dye content) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...