1,3,5-Triformylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,5-benzenetricarbaldehyde, Trimesic formaldehyde, Benzene-1,3,5-tricarbaldehyde

44655-0010 44655-0050
TS44655-0010EA 250 USD
TS44655-0010 TS44655-0050
1,3,5-Triformylbenzene 98%
1,3,5-Triformylbenzene
Formula: C₉H₆O₃
MW: 162.14 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00182470
CAS Number: 3163-76-6

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR