Ethynylmagnesium bromide 0.5 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

43912-1000 43912-8000
TS43912-1000EA 75 USD
TS43912-1000 TS43912-8000
Ethynylmagnesium bromide 0.5 M in THF, AcroSeal™
Ethynylmagnesium bromide
Formula: C₂HBrMg
Density: 0.94 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00075342
CAS Number: 4301-14-8
UN: 3399
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Colorless to yellow to gray to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR