Methyl cis-11-eicosenoate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: cis-11-Eicosenoic acid methyl ester, methyl (Z)-icos-11-enoate, (Z)-Methyl icos-11-enoate, (11Z)-11-Icosenoic acid methyl ester

42853-0010 42853-0050
TS42853-0010EA 412 USD
TS42853-0010 TS42853-0050
Methyl cis-11-eicosenoate 99%
Methyl cis-11-eicosenoate
Formula: C₂₁H₄₀O₂
MW: 324.549 g/mol
Melting Pt: –15 °C
Flash Pt: >113 °C
Storage Temperature: Freezer
MDL Number: MFCD00010458
CAS Number: 2390-09-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR