Ammonium trifluoromethanesulfonate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methanesulphonic acid,1,1,1-trifluoro-, ammonium salt (1:1), Ammonium triflate
38873-0250
38873-0500
AC38873-0050
TS38873-0250EA
179
USD
TS38873-0250
TS38873-0500
TS38873-0050
Ammonium trifluoromethanesulfonate 99%
Ammonium trifluoromethanesulfonate
Formula:
CH₄F₃NO₃S MW: 167.102 g/mol Melting Pt: 225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075332 CAS Number: 38542-94-8 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Elemental Analysis | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...