Iron(III) sulfate pentahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: ferric sulphate hexahydrate, ferric sulphate, ferric sulphate heptahydrate

34523-1000 34523-5000 AC34523-0050 AC34523-0051
TS34523-1000EA 54 USD
TS34523-1000 TS34523-5000 TS34523-0050 TS34523-0051
Iron(III) sulfate pentahydrate 97%
Iron(III) sulfate pentahydrate
Formula: Fe₂H₁₀O₁₇S₃
MW: 489.933 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149716
CAS Number: 142906-29-4
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Appearance Off-white to yellow to beige to gray Crystalline or granular powder
Degree of hydration May vary
Titration with Na2S2O3 20.7 to 23 % (as Fe)
Chloride (Cl) ≤50 ppm
Nitrate (NO3) ≤200 ppm
Phosphate (PO4) ≤30 ppm
Arsenic (As) ≤5 ppm
Copper (Cu) ≤20 ppm
Zinc (Zn) ≤50 ppm
Ferrous iron (Fe 2+) ≤200 ppm
Insoluble matter ≤0.01 % (in HCl)
Subst not pptd by NH3 ≤0.05 % (as SO4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR