1-Pentanesulfonic acid, sodium salt hydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
27037-1000
AC27037-0050
AC27037-0250
TS27037-1000EA
497
USD
TS27037-1000
TS27037-0050
TS27037-0250
1-Pentanesulfonic acid, sodium salt hydrate 99%
1-Pentanesulphonic acid sodium salt hydrate
Formula:
C₅H₁₁NaO₃S·xH₂O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149548 CAS Number: 1266615-65-9 EINECS: 245-208-4 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤9.6 % (K.F.) |
Solubility | (10 % in water) Clear colorless |
Titration after Ion exchange | 98.5 to 101.5 % (On dried substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...