Lithium tetrakis(pentafluorophenyl)borate ethyl etherate

Supplier: Thermo Scientific Chemicals
Warning

45853-0010 AC45853-1000
TS45853-0010EA 310 USD
TS45853-0010 TS45853-1000
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
Formula: C₂₄BF₂₀Li·2.5C₄H₁₀O
MW: 871.28 g/mol
Melting Pt: 117…122 °C
Storage Temperature: Ambient
CAS Number: 371162-53-7

Order Now

Specification Test Results

Appearance White to light beige Powder or crystals
Melting point 117 to 122°C
NMR (19F-NMR) ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR