Isopropylmagnesium chloride - LiCl complex 1.3 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Isopropylmagnesium chloride - Lithium chloride complex, Turbo Grignard

38628-1000 AC38628-8000
TS38628-1000EA 117 USD
TS38628-1000 TS38628-8000
Isopropylmagnesium chloride - LiCl complex 1.3 M in THF, AcroSeal™
Isopropylmagnesium chloride - LiCl complex
Formula: C₃H₇ClMg·LiCl
MW: 145.24 g/mol
Density: 0.98 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD07784514
CAS Number: 807329-97-1
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear yellow to gray to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.1 to 1.5 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR