Hydroxy naphthol blue disodium salt ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydroxynaphthol blue sodium salt, Hydroxy napthol blue, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulpho-3-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate

20488-1000 20488-5000 20488-0050
TS20488-1000EA 155 USD
TS20488-1000 TS20488-5000 TS20488-0050
Hydroxy naphthol blue disodium salt ACS
Hydroxy naphthol blue disodium salt
Formula: C₂₀H₁₂N₂O₁₁S₃·2Na
MW: 598.479 g/mol
Melting Pt: 274 °C
Storage Temperature: Ambient
MDL Number: MFCD00004075
CAS Number: 165660-27-5

Order Now

Specification Test Results

Appearance Blue to dark purple Crystalline powder or crystals
Infrared spectrum Conforms
Suitability (for Ca determination) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR