Methyl viologen dichloride hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

22732-0010 22732-0050
TS22732-0010EA 83 USD
TS22732-0010 TS22732-0050
Methyl viologen dichloride hydrate 98%
Methyl viologen dichloride hydrate
Formula: C₁₂H₁₄Cl₂N₂·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150001
CAS Number: 75365-73-0
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Beige to pink to brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥97.5 % (on dry substance)
Water ≤18 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR