DL-Isocitric acid, trisodium salt hydrate 95%
Supplier: Thermo Scientific Chemicals
20501-0010
20501-0050
20501-0250
TS20501-0010EA
78
USD
TS20501-0010
TS20501-0050
TS20501-0250
DL-Isocitric acid, trisodium salt hydrate 95%
DL-Isocitric acid, trisodium salt hydrate
Formula:
C₆H₅Na₃O₇·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149284 CAS Number: 1637-73-6 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤20 % (K.F.) |
Titration with HClO4 | ≥94 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...