potassium ethenyl sulfate, M.W. ∼ 175,000
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Polyvinyl sulphate potassium salt
41814-0050
41814-0250
41814-1000
TS41814-0050EA
226
USD
TS41814-0050
TS41814-0250
TS41814-1000
potassium ethenyl sulfate, M.W. ∼ 175,000
Polyvinylsulfuric acid potassium salt
Formula:
(C₂H₃KO₄S)n Storage Temperature: Refrigerator |
MDL Number:
MFCD00081923 CAS Number: 26837-42-3 |
Specification Test Results
Appearance | Off-white to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥21.2 % (K) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...