Hydrazine acetate 97%

Supplier: Thermo Scientific Chemicals
Danger

41175-0250 41175-0050
TS41175-0250EA 246 USD
TS41175-0250 TS41175-0050
Hydrazine acetate 97%
Hydrazine acetate
Formula: H₂NNH₂·CH₃CO₂H
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00013141
CAS Number: 7335-65-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Adhering powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR