2-(Trimethylsilylmethyl)allyl acetate 95%

Supplier: Thermo Scientific Chemicals
Warning

38138-0050 AC38138-0010 AC38138-0250
TS38138-0050EA 266 USD
TS38138-0050 TS38138-0010 TS38138-0250
2-(Trimethylsilylmethyl)allyl acetate 95%
2-(Trimethylsilylmethyl)allyl acetate
Formula: C₉H₁₈O₂Si
Boiling Pt: 95 °C (7 mmHg)
Density: 0.877 g/cm³
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075170
CAS Number: 72047-94-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4420 (20°C, 589 nm)
GC ≥92 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR