(Ethylthio)trimethylsilane 90%

Supplier: Thermo Scientific Chemicals
Danger

34500-0100 AC34500-0010
TS34500-0100EA 232 USD
TS34500-0100 TS34500-0010
(Ethylthio)trimethylsilane 90%
(Ethylthio)trimethylsilane
Formula: C₅H₁₄SSi
Boiling Pt: 69…70 °C (100 mmHg)
Density: 0.83 g/cm³
Flash Pt: 16 °C
Storage Temperature: Ambient
CAS Number: 5573-62-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4510 (20°C, 589 nm)
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR