3-Chloroperoxybenzoic acid 70-75% in water balance 3-Chlorobenzoic acid and water
Supplier: Thermo Scientific Chemicals
|
Danger
|
25579-0010
25579-1000
25579-0250
25579-5000
TS25579-0010EA
618
USD
TS25579-0010
TS25579-1000
TS25579-0250
TS25579-5000
3-Chloroperoxybenzoic acid 70-75% in water balance 3-Chlorobenzoic acid and water
3-Chloroperoxybenzoic acid
Formula:
C₇H₅ClO₃ Melting Pt: 92…94 °C Density: 0.56 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00002127 CAS Number: 937-14-4 UN: 3106 ADR: 5.2, |
Specification Test Results
Appearance | White Moist powder |
Titration Iodometric | 70 to 75 % |
Water | % balance (K.F.) |
HPLC | di-m-Chlorobenzoyl peroxide: < 0.1% Typical |
3-Chlorobenzoic acid | ≤12 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...