Methyllithium 1.6 M in diethyl ether (± 5% w/v), AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyllithium - lithium bromide complex, Methyllithium solution
18875-1000
AC18875-8000
TS18875-1000EA
83
USD
TS18875-1000
TS18875-8000
Methyllithium 1.6 M in diethyl ether (± 5% w/v), AcroSeal™
Methyllithium
Formula:
CH₃Li MW: 21.975 g/mol Density: 0.7 g/cm³ Flash Pt: –17 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008253 CAS Number: 917-54-4 UN: 3394 ADR: 4.2, |
Specification Test Results
Appearance | Colorless to yellow Solution (May develop some turbidity or precipitate) |
Titration Total base | 1.43 to 1.75 M |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...