Sodium hexacyanoferrate(II) decahydrate

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium ferrocyanide decahydrate

45929-0010 AC45929-0050 AC45929-2500
TS45929-0010EA 57 USD
TS45929-0010 TS45929-0050 TS45929-2500
Sodium hexacyanoferrate(II) decahydrate
Sodium hexacyanoferrate(II) decahydrate
Formula: C₆H₂₀FeN₆Na₄O₁₀
MW: 484.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085360
CAS Number: 14434-22-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Loss on drying ≤1 % (free moisture)
Titration ≥98.5 %
Chloride (Cl) ≤0.1 %
Sulfate (SO4) ≤0.07 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR