Nile red 99%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one

41571-0010 41571-1000
TS41571-0010EA 637 USD
TS41571-0010 TS41571-1000
Nile red 99%, pure
Nile red
Formula: C₂₀H₁₈N₂O₂
MW: 318.38 g/mol
Melting Pt: 203…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00011639
CAS Number: 7385-67-3

Order Now

Specification Test Results

Appearance Dark green to dark red to brown Powder
Infrared spectrum Conforms
UV spectrum Complies
UV spectrophotometry ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR