Nicotine ditartrate dihydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

41566-0100 41566-0500
TS41566-0100EA 90 USD
TS41566-0100 TS41566-0500
Nicotine ditartrate dihydrate 98%
Nicotine ditartrate dihydrate
Formula: C₁₀H₁₄N₂·2C₄H₆O₆·2H₂O
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00069368
CAS Number: 6019-06-3
UN: 1659
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water 5 to 10 % (K.F.)
Titration with TBAH ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR