Methylenediphenyl-4,4'-diisocyanate 98%, flakes

Supplier: Thermo Scientific Chemicals
Danger

41428-5000 41428-1000
TS41428-5000EA 79.2 USD
TS41428-5000 TS41428-1000
Methylenediphenyl-4,4'-diisocyanate 98%, flakes
Methylenediphenyl-4,4'-diisocyanate
Formula: C₁₅H₁₀N₂O₂
Boiling Pt: 392 °C (760 mmHg)
Melting Pt: 40 °C
Density: 1.22 g/cm³
Flash Pt: 202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036131
CAS Number: 101-68-8

Order Now

Specification Test Results

Appearance White to yellow Flakes
Infrared spectrum Conforms
Melting point 39 to 44 °C
Titration ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR