Isopropylmagnesium chloride 2.0 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: i-Propylmagnesium chloride

21285-1000 AC21285-8000
TS21285-1000EA 64 USD
TS21285-1000 TS21285-8000
Isopropylmagnesium chloride 2.0 M in THF, AcroSeal™
Isopropylmagnesium chloride
Formula: C₃H₇ClMg
MW: 102.84 g/mol
Boiling Pt: 62.4 °C (899.8hPa)
Melting Pt: 5 °C
Density: 0.97 g/cm³
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00000466
CAS Number: 1068-55-9
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Colorless to yellow to gray to dark brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.80 to 2.20 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR