Isohexane for pesticide residue analysis contains <5% n-Hexane

Supplier: Thermo Scientific Chemicals
Danger

38382-0025 383820010
TS38382-0025EA 184 USD
TS38382-0025 TS383820010
Isohexane for pesticide residue analysis contains <5% n-Hexane
Isohexane (mixture of isomers)
Formula: C₆H₁₄
Boiling Pt: 60 °C (760 mmHg)
Melting Pt: –154 °C
Density: 0.66 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 73513-42-5
EINECS: 295-570-2
UN: 1208
ADR: 3,II
REACH: 01-2119484651-34

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.01 % (Coulometric)
GC (ECD) Passes test, (NPD) Passes test, (FID) Passes test
Residue after evaporation ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR