Hydroxychloroquine sulfate 98%

Supplier: Thermo Scientific Chemicals
Warning

26301-0250 26301-1000
TS26301-0250EA 231 USD
TS26301-0250 TS26301-1000
Hydroxychloroquine sulfate 98%
Hydroxychloroquine sulfate
Formula: C₁₈H₂₆ClN₃O
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00078203
CAS Number: 747-36-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 % (105°C, 2 h)
Heavy metals ≤20 ppm
Sulfated ash ≤0.2 %
UV spectrophotometry ≥97.5 % (on dried basis)
Chloride (Cl) ≤350 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR