Fluorescamine, pure

Supplier: Thermo Scientific Chemicals

19167-0025 191670250 191671000 191675000
TS19167-0025EA 1077 USD
TS19167-0025 TS191670250 TS191671000 TS191675000
Fluorescamine, pure
Fluorescamine
Formula: C₁₇H₁₀O₄
Melting Pt: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00005928
CAS Number: 38183-12-9

Order Now

Specification Test Results

Appearance Light yellow Powder
Infrared spectrum Conforms
Melting point 155 to 157 °C
Specific extinction E (1%/1cm) ≥900 (235 nm, ether)
Suitability fluorescence ind. for prim. amines and amino acids


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR