Dimethyl(2-hydroxyethyl)phosphonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2-Hydroxyethyl)phosphonic Acid Dimethyl Ester

33816-0250 33816-0050
TS33816-0250EA 445 USD
TS33816-0250 TS33816-0050
Dimethyl(2-hydroxyethyl)phosphonate 98%
Dimethyl(2-hydroxyethyl)phosphonate
Formula: C₄H₁₁O₄P
MW: 154.10 g/mol
Boiling Pt: 73 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.7 g/cm³
Flash Pt: –16 °C
Storage Temperature: Ambient
MDL Number: MFCD00043376
CAS Number: 54731-72-5
EINECS: 209-263-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4130 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR