Allylmagnesium bromide 1 M in diethyl ether, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

20953-1000 AC20953-8000
TS20953-1000EA 87 USD
TS20953-1000 TS20953-8000
Allylmagnesium bromide 1 M in diethyl ether, AcroSeal™
Allylmagnesium bromide
Formula: C₃H₅BrMg
Density: 0.851 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00000044
CAS Number: 1730-25-2
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Yellow-green to gray Solution
Titration Grignard 0.9 to 1.1 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR