Quinoline yellow sulfonic acid sodium salt (Acid yellow 3) 95%, water soluble, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
D and C Yellow No. 10, Quinoline Yellow S, Quinaldine Yellow, Food yellow 13, CI 47005
27734-0250
27734-1000
TS27734-0250EA
79
USD
TS27734-0250
TS27734-1000
Quinoline yellow sulfonic acid sodium salt (Acid yellow 3) 95%, water soluble, pure
Quinoline yellow sulfonic acid sodium salt (Acid yellow 3)
Formula:
C₁₈H₁₁NO₂.xNaO₃S Storage Temperature: Ambient |
MDL Number:
MFCD00080727 CAS Number: 8004-92-0 |
Specification Test Results
Appearance | Yellow to orange-brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...