6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trolox, 6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid

21894-0010 21894-0050 21894-0250
TS21894-0010EA 85 USD
TS21894-0010 TS21894-0050 TS21894-0250
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid 97%
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid
Formula: C₁₄H₁₈O₄
MW: 250.29 g/mol
Melting Pt: 187…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00006846
CAS Number: 53188-07-1

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 194 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR