4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2,2,6,6-Pentamethylpiperidin-4-ol, 1,2,2,6,6-Pentamethyl-4-piperidinol

30086-0010 30086-2500
TS30086-0010EA 311 USD
TS30086-0010 TS30086-2500
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 99%
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine
Formula: C₁₀H₂₁NO
MW: 171.28 g/mol
Boiling Pt: 238 °C (760 mmHg)
Melting Pt: 70…76 °C
Density: 0.967 g/cm³
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00191220
CAS Number: 2403-89-6
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow Flakes
Infrared spectrum Conforms
Melting point 70 to 76 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR