4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) mixture of isomers 99% (mixture of isomers)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(4-amino-3-methylcyclohexyl)methane, 4,4'-Methylenebis(2-methylcyclohexylamine), 4,4'-Methylenebis(1-amino-2-methylcyclohexane), 4,4'-Diamino-3,3'-dimethyldicyclohexylmethane

45024-0010 450242500
TS45024-0010EA 270 USD
TS45024-0010 TS450242500
4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) mixture of isomers 99% (mixture of isomers)
4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers)
Formula: C₁₅H₃₀N₂
Boiling Pt: 93…100 °C (760 mmHg)
Density: 0.94 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075535
CAS Number: 6864-37-5
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5000 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR