4-(Chloromethyl)styrene 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Chloromethyl)-4-vinylbenzene, 4-Vinylbenzyl Chloride

42252-5000 422520250 422521000
TS42252-5000EA 674 USD
TS42252-5000 TS422520250 TS422521000
4-(Chloromethyl)styrene 90% tech.
4-(Chloromethyl)styrene
Formula: C₉H₉Cl
MW: 152.62 g/mol
Density: 1.08 g/cm³
Flash Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051362
CAS Number: 1592-20-7
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to dark yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥88 %
Stabilizer About 500 ppm Nitrocresol + Nitrophenol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR