Polyvinyl alcohol, M.W. ∼ 145,000-180,000 88%, hydrolysed

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

45015-0010 450150250 450155000
TS45015-0010EA 152 USD
TS45015-0010 TS450150250 TS450155000
Polyvinyl alcohol, M.W. ∼ 145,000-180,000 88%, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 230…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance White to off-white Granular solid
Infrared spectrum Conforms
Viscosity 45 to 55 cP (4 % in water at 20°C)
Degree of hydrolysis 87 to 89 mol %
Ash ≤5 % (Na2O)
pH 4.5 to 6.5 (4 % in water)
Total volatile ≤5 % (including water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR