2,3-Naphthalenedialdehyde (NDA) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Naphthalene-2,3-dicarbaldehyde, Naphthalene-2,3-dicarboxaldehyde, NDA
29472-5000
29472-1000
TS29472-5000EA
498
USD
TS29472-5000
TS29472-1000
2,3-Naphthalenedialdehyde (NDA) 95%
2,3-Naphthalenedialdehyde (NDA)
Formula:
C₁₂H₈O₂ Melting Pt: 134…135 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016618 CAS Number: 7149-49-7 |
Specification Test Results
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...