(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 97%

Supplier: Thermo Scientific Chemicals
Warning

29451-0050 29451-0010 29451-0250
TS29451-0050EA 287 USD
TS29451-0050 TS29451-0010 TS29451-0250
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 97%
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt
Formula: C₈H₁₈N₂O₄S
Melting Pt: 76…79 °C
Storage Temperature: Freezer
MDL Number: MFCD00077815
CAS Number: 29684-56-8

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR